Learn More
BioVision IncSupplier Diversity Partner MTS Cell Proliferation Colorimetric Assay Kit: MTS Cell Proliferation Colorimetric Assay Kit: Faster & Sensitive MTS based Assay to Measure Cell Proliferation in response to growth factors, cytokines, mitogens and nutrients etc. 500 & 2500 Assays. DFS Item
Manufacturer:  BioVision IncSupplier Diversity Partner K300500
MTS Cell Proliferation Colorimetric Assay Kit: BioVisions MTS Cell Proliferation Assay Kit is a colorimetric method for sensitive quantification of viable cells in proliferation and cytotoxicity assay. The method is based on the reduction of MTS tetrazolium compound by viable cells to generate a colored formazan product that is soluble in cell culture media. Viable cells can be quantified by measuring the absorbance at 490-500 nm. The assay can be used for the measurement of cell proliferation in response to growth factors, cytokines, mitogens, etc. It can also be used for the analysis of cytotoxic compounds like anticancer drugs and many other toxic agents and pharmaceutical compounds. BioVisions MTS assay is performed by adding the reagent directly into the cell culture media without the intermittent steps which are required in the routine MTT assay. In addition, this high-throughput assay requires no washing or solubilization step and can be performed in 96-well microtiter plate
Catalog No. 50-111-9675