Mystaire™ Aura™ Elite Series Ductless Fume Hood Accessory, Gas Phase Filters
Provides operator safety from potentially harmful fumes and odors
Manufacturer: Mystaire™ MY5814
 View more versions of this product
Catalog No. 15-338-433
  Check Availability
Add to Cart
    Specifications
    For Use With (Equipment) Safety for Aura Elite 30 and 54 Ductless Fume Hood
    Product Type Gas Phase Filter
    Filter Type Carbon
    For Use With (Application) Minerals and acids
  • Description & Specifications
Related Products
Product Content Feedback