Learn More
Mystaire™ Room Air Purifier and Ductless Fume Extractor Main Filter
For use with the Mystaire Room Air Purifier (FE-1200), Single-Arm Ductless Fume Extractor (FE-1200A) and Dual-Arm Ductless Fume Extractor (FE-1200B).
Supplier: Mystaire™ FE1007
Description
- Made in the U.S.A.
- Each filter application is reviewed to ensure a proper filtration package is selected
- Filter performance verified by Fourier Transform Infrared Spectroscopy (FTIR)
Specifications
FE-1200, FE-1200 A and FE-1200 B | |
Filter | |
Gas Phase Carbon | |
Formaldehyde |