Learn More
Mystaire™ Aura™ Elite Series Ductless Fume Hood Gas Phase Filters
For Aura Elite Series 30, 42 and 54 in. Ductless Fume Hoods. Provides operator safety from potentially harmful fumes and odors.
Manufacturer:  Mystaire™ MY5904
 View more versions of this product
Catalog No. 15-338-424
Add to cart
Description
Description
  • Manufactured with high-grade coconut shell carbon
  • Activated carbon filter that provides more surface area for a higher degree of absorbency
  • Chemically impregnated carbon for increased load capacity
Specifications
Specifications
Gas Phase Filter
Carbon
Hydrocarbons
Safety for Aura Elite 42 Ductless Fume Hood