Learn More
Mystaire™ Aura™ Elite Series Ductless Fume Hood Gas Phase Filters
SDP
For Aura Elite Series 30, 42 and 54 in. Ductless Fume Hoods. Provides operator safety from potentially harmful fumes and odors.
Supplier:  Mystaire™ MY3808
 View more versions of this product
Catalog No. 15-338-445
Add to Cart
Description
Description
  • Manufactured with high-grade coconut shell carbon
  • Activated carbon filter that provides more surface area for a higher degree of absorbency
  • Chemically impregnated carbon for increased load capacity
Specifications
Specifications
Carbon
Aura Elite 30 and 54 Ductless Fume Hood
Gas Phase Filter
Ammonia, Amines
Product Suggestions
Product Suggestions