Learn More
Mystaire™ Aura™ Elite Series Ductless Fume Hood Gas Phase Filters
For Aura Elite Series 30, 42 and 54 in. Ductless Fume Hoods. Provides operator safety from potentially harmful fumes and odors.
Manufacturer:  Mystaire™ MY3816
 View more versions of this product
Catalog No. 15-338-451
Add to cart
Description
Description
  • Manufactured with high-grade coconut shell carbon
  • Activated carbon filter that provides more surface area for a higher degree of absorbency
  • Chemically impregnated carbon for increased load capacity
Specifications
Specifications
Carbon
Multi-Purpose
Aura Elite 30 and 54 Ductless Fume Hood
Gas Phase Filter