Learn More
Mystaire™ Aura™ Elite Series Ductless Fume Hood Gas Phase Filters
For Aura Elite Series 30, 42 and 54 in. Ductless Fume Hoods. Provides operator safety from potentially harmful fumes and odors.
$316.00 - $906.00
Specifications
Filter Type Carbon
Product Type Gas Phase Filter
View More Specs
Products 26
Catalog Number Mfr. No. For Use With (Application) For Use With (Equipment) Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Application) For Use With (Equipment) Price Quantity & Availability  
15-338-440
View Documents
Mystaire™
MY2804
Hydrocarbons Aura Elite 42 Ductless Fume Hood
Each for $484.00
Add to cart
15-338-441
View Documents
Mystaire™
MY3804
Hydrocarbons Aura Elite 30 and 54 Ductless Fume Hood
Each for $815.00
Add to cart
15-338-442
View Documents
Mystaire™
MY2807
Formaldehyde Aura Elite 42 Ductless Fume Hood
Each for $639.00
Add to cart
15-338-443
View Documents
Mystaire™
MY3807
Formaldehyde Aura Elite 30 and 54 Ductless Fume Hood
Each for $786.00
Add to cart
15-338-444
View Documents
Mystaire™
MY2808
Ammonia and Amines Aura Elite 42 Ductless Fume Hood
Each for $639.00
Add to cart
15-338-445
View Documents
Mystaire™
MY3808
Ammonia and Amines Aura Elite 30 and 54 Ductless Fume Hood
Each for $786.00
Add to cart
15-338-446
View Documents
Mystaire™
MY2809
Sulphur Aura Elite 42 Ductless Fume Hood
Each for $643.00
Add to cart
15-338-447
View Documents
Mystaire™
MY3809
Sulphur Aura Elite 30 and 54 Ductless Fume Hood
Each for $774.00
Add to cart
15-338-448
View Documents
Mystaire™
MY2814
Mineral Acids Aura Elite 42 Ductless Fume Hood
Each for $639.00
Add to cart
15-338-449
View Documents
Mystaire™
MY3814
Mineral Acids Aura Elite 30 and 42 Ductless Fume Hood
Each for $741.00
Add to cart
15-338-450
View Documents
Mystaire™
MY2816
Multi-Purpose Aura Elite 42 Ductless Fume Hood
Each for $781.00
Add to cart
15-338-451
View Documents
Mystaire™
MY3816
Multi-Purpose Aura Elite 30 and 54 Ductless Fume Hood
Each for $814.00
Add to cart
15-338-452
View Documents
Mystaire™
MY2817
Custom Blend Aura Elite 42 Ductless Fume Hood
Each for $770.00
Add to cart
15-338-453
View Documents
Mystaire™
MY3817
Custom Blend Aura Elite 30 and 54 Ductless Fume Hood
Each for $906.00
Add to cart
15-338-424
View Documents
Mystaire™
MY5904
Hydrocarbons Safety for Aura Elite 42 Ductless Fume Hood
Each for $317.00
Add to cart
15-338-425
View Documents
Mystaire™
MY5804
Hydrocarbons Safety for Aura Elite 30 and 54 Ductless Fume Hood
Each for $316.00
Add to cart
15-338-426
View Documents
Mystaire™
MY5907
Formaldehyde Safety for Aura Elite 42 Ductless Fume Hood
Each for $368.00
Add to cart
15-338-427
View Documents
Mystaire™
MY5807
Formaldehyde Safety for Aura Elite 30 and 54 Ductless Fume Hood
Each for $428.00
Add to cart
15-338-428
View Documents
Mystaire™
MY5908
Ammonia and Amines Safety for Aura Elite 42 Ductless Fume Hood
Each for $419.00
Add to cart
15-338-429
View Documents
Mystaire™
MY5808
Ammonia and Amines Safety for Aura Elite 30 and 54 Ductless Fume Hood
Each for $420.00
Add to cart
15-338-430
View Documents
Mystaire™
MY5909
Sulphur Safety for Aura Elite 42 Ductless Fume Hood
Each for $435.00
Add to cart
15-338-431
View Documents
Mystaire™
MY5809
Sulphur Safety for Aura Elite 30 and 54 Ductless Fume Hood
Each for $388.00
Add to cart
15-338-432
View Documents
Mystaire™
MY5914
Minerals and Acids Safety for Aura Elite 42 Ductless Fume Hood
Each for $369.00
Add to cart
15-338-433
View Documents
Mystaire™
MY5814
Minerals and Acids Safety for Aura Elite 30 and 54 Ductless Fume Hood
Each for $393.00
Add to cart
15-338-434
View Documents
Mystaire™
MY5917
Custom Blend Safety for Aura Elite 42 Ductless Fume Hood
Each for $496.00
Add to cart
15-338-435
View Documents
Mystaire™
MY5817
Custom Blend Safety for Aura Elite 30 and 54 Ductless Fume Hood
Each for $492.00
Add to cart
Description
Description
  • Manufactured with high-grade coconut shell carbon
  • Activated carbon filter that provides more surface area for a higher degree of absorbency
  • Chemically impregnated carbon for increased load capacity
Specifications
Specifications
Carbon
Gas Phase Filter