Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MyT1L Rabbit anti-Mouse, Unconjugated, Polyclonal, MilliporeSigma™

Rabbit Polyclonal Antibody

$147.00 - $372.00

Specifications

Antigen MyT1L
Applications ChIP assay, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Conjugate Unconjugated
Format Affinity Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ABE291510MI
View Documents
MilliporeSigma
ABE2915100UG
100 μg
Each for $372.00
Add to cart
 
ABE291525MI
View Documents
MilliporeSigma
ABE291525UG
25 μg
Each for $147.00
Add to cart
 
Description

Description

MyT1L Polyclonal antibody specifically detects MyT1L in Mouse samples. It is validated for ChIP-seq, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blotting.
Specifications

Specifications

MyT1L
Unconjugated
Rabbit
Myelin transcription factor 1-like protein;MyT1-L;Neural zinc finger factor 1;NZF-1;Postmitotic neural gene 1 protein;Zinc finger protein Png-1
His-tagged recombinant fragment corresponding to 250 amino acids from the N-terminal half of mouse Myelin transcription factor 1-like protein
Primary
Stable for 1 year at -20°C from date of receipt.
ChIP assay, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Affinity Purified
Mouse
Myt1l, Kiaa1106, Nzf1, Png1
Polyclonal
Affinity Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit