Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N,1-Di-Fmoc-L-histidine, 95%

Click to view available options
Quantity:
1g
5g
25g
98929-98-7
C36H29N3O6
599.643
MFCD00070658
WTBXFPFCUZKREB-XIFFEERXSA-N
fmoc-his fmoc-oh,n,n'-bis 9-fluorenylmethyloxycarbonyl-l-histidine,s-3-1-9h-fluoren-9-yl methoxy carbonyl-1h-imidazol-4-yl-2-9h-fluoren-9-yl methoxy carbonyl amino propanoic acid,l-histidine, n,1-bis 9h-fluoren-9-ylmethoxy carbonyl,fmoc-his fmoc,2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-1-9h-fluoren-9-ylmethoxy carbonyl imidazol-4-yl propanoic acid,ambotzfaa1497,fmoc-l-his fmoc-oh,pubchem10020,fmoc-histidine fmoc-oh
45072174
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[1-(9H-fluoren-9-ylmethoxycarbonyl)imidazol-4-yl]propanoic acid
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CN(C=N4)C(=O)OCC5C6=CC=CC=C6C7=CC=CC=C57)C(=O)O
98929-98-7
C36H29N3O6
599.643
MFCD00070658
WTBXFPFCUZKREB-XIFFEERXSA-N
fmoc-his fmoc-oh,n,n'-bis 9-fluorenylmethyloxycarbonyl-l-histidine,s-3-1-9h-fluoren-9-yl methoxy carbonyl-1h-imidazol-4-yl-2-9h-fluoren-9-yl methoxy carbonyl amino propanoic acid,l-histidine, n,1-bis 9h-fluoren-9-ylmethoxy carbonyl,fmoc-his fmoc,2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-1-9h-fluoren-9-ylmethoxy carbonyl imidazol-4-yl propanoic acid,ambotzfaa1497,fmoc-l-his fmoc-oh,pubchem10020,fmoc-histidine fmoc-oh
45072174
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[1-(9H-fluoren-9-ylmethoxycarbonyl)imidazol-4-yl]propanoic acid
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CN(C=N4)C(=O)OCC5C6=CC=CC=C6C7=CC=CC=C57)C(=O)O
Quantity 5g
Formula Weight 599.63
Percent Purity 95%
Chemical Name or Material N,1-Di-Fmoc-L-histidine

TSCA : No

Recommended Storage : Store at -20°C