Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-(1-Glycyl-L-prolyl)alanine, 97%

Click to view available options
Quantity:
250mg
1g
837-83-2
C10H19N3O5
261.278
MFCD00037344
RLLPBIPOWCFAOP-LEUCUCNGSA-N
H-Gly-Pro-Ala-OH; Glycyl-L-prolyl-L-alanine
91872902
(2S)-2-[[(2S)-1-(2-aminoacetyl)pyrrolidine-2-carbonyl]amino]propanoic acid;hydrate
CC(C(=O)O)NC(=O)C1CCCN1C(=O)CN.O
837-83-2
C10H19N3O5
261.278
MFCD00037344
RLLPBIPOWCFAOP-LEUCUCNGSA-N
H-Gly-Pro-Ala-OH; Glycyl-L-prolyl-L-alanine
91872902
(2S)-2-[[(2S)-1-(2-aminoacetyl)pyrrolidine-2-carbonyl]amino]propanoic acid;hydrate
CC(C(=O)O)NC(=O)C1CCCN1C(=O)CN.O
Beilstein 5288874
Formula Weight 243.26
Physical Form Powder
Percent Purity 97%
Color White
Quantity 1g
Chemical Name or Material N-(1-Glycyl-L-prolyl)alanine

storageNote1 : -30° to -10°C