Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(1-Naphthyl)-Ethylenediamine Dihydrochloride, Reagents

Catalog No. 01337160
Click to view available options
Quantity:
10 g
25 g

N-(1-Naphthyl)-Ethylenediamine Dihydrochloride, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 1465-25-4
Molecular Formula C12H16Cl2N2
Molecular Weight (g/mol) 259.17
InChI Key MZNYWPRCVDMOJG-UHFFFAOYSA-N
Synonym N1-(Naphthalen-1-yl)ethane-1,2-diamine dihydrochloride
IUPAC Name dihydrogen N1-(naphthalen-1-yl)ethane-1,2-diamine dichloride
SMILES [H+].[H+].[Cl-].[Cl-].NCCNC1=C2C=CC=CC2=CC=C1

Specifications

Boiling Range 371°C
Color White to Pink
Boiling Point 371°C
Packaging Glass Bottle
Quantity 25 g
Specific Gravity 1.1
Grade Reagent
Physical Form Solid
Chemical Name or Material N-(1-Naphthyl)-Ethylenediamine <br />Dihydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.