Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(1-Naphthyl)-N-(phenylmethylene)amine, 97%, Maybridge

Click to view available options
Quantity:
1g
10g
Packaging:
Amber Glass Bottle
890-51-7
C17H13N
231.298
MFCD02235774
NQPYSYDZNCHIQY-UHFFFAOYSA-N
n-1-naphthyl-n-phenylmethylene amine,n-benzylidenenaphthalen-1-amine,n-e-phenylmethylidene naphthalen-1-amine,1z-1-naphthyl-2-phenyl-1-azaethene,n-naphthalen-1-yl-1-phenylmethanimine,1z-n-naphthalen-1-yl-1-phenylmethanimine,n-benzylidene-1-napthylamine,benzylidene-a-napthylamine,maybridge3_000265,n-benzylidene-1-naphthalenamine
1201463
N-naphthalen-1-yl-1-phenylmethanimine
C1=CC=C(C=C1)C=NC2=CC=CC3=CC=CC=C32
890-51-7
C17H13N
231.298
MFCD02235774
NQPYSYDZNCHIQY-UHFFFAOYSA-N
n-1-naphthyl-n-phenylmethylene amine,n-benzylidenenaphthalen-1-amine,n-e-phenylmethylidene naphthalen-1-amine,1z-1-naphthyl-2-phenyl-1-azaethene,n-naphthalen-1-yl-1-phenylmethanimine,1z-n-naphthalen-1-yl-1-phenylmethanimine,n-benzylidene-1-napthylamine,benzylidene-a-napthylamine,maybridge3_000265,n-benzylidene-1-naphthalenamine
1201463
N-naphthalen-1-yl-1-phenylmethanimine
C1=CC=C(C=C1)C=NC2=CC=CC3=CC=CC=C32
Quantity 10g
Packaging Amber Glass Bottle
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 231.3
Chemical Name or Material N-(1-naphthyl)-N-(phenylmethylene)amine