Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-1-Naphthylethylenediamine Dihydrochloride, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
100g
25g
Packaging:
Wide Mouth Amber Glass Bottle

Chemical Identifiers

1465-25-4
C12H16Cl2N2
259.174
MZNYWPRCVDMOJG-UHFFFAOYSA-N
15106
CHEBI:53452

High quality chemicals for laboratory and specialized industrial use

    For Nitrogen Dioxide Determination (ASTM D-1607)
CAS 1465-25-4
Packaging Wide Mouth Amber Glass Bottle
Molecular Formula C12H16Cl2N2
Quantity 25g
InChI Key MZNYWPRCVDMOJG-UHFFFAOYSA-N
SMILES C1=CC=C2C(=C1)C=CC=C2NCCN.Cl.Cl
IUPAC Name N'-naphthalen-1-ylethane-1,2-diamine;dihydrochloride
Molecular Weight (g/mol) 259.174
PubChem CID 15106
ChEBI CHEBI:53452
Formula Weight 259.18
Chemical Name or Material N-1-Naphthylethylenediamine Dihydrochloride
Grade Baker Analyzed™ ACS Reagent
Show More Show Less