Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-1-Naphthylethylenediamine Dihydrochloride, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
25g
100g
Packaging:
Wide Mouth Amber Glass Bottle
1465-25-4
C12H16Cl2N2
259.174
MZNYWPRCVDMOJG-UHFFFAOYSA-N
n-1-naphthyl ethylenediamine dihydrochloride,n1-naphthalen-1-yl ethane-1,2-diamine dihydrochloride,marshall's reagent,n-1-naphthylethylenediamine dihydrochloride,ccris 425,unii-h734599kjl,1,2-ethanediamine, n-1-naphthalenyl-, dihydrochloride,2-1-naphthylamino ethylamine 2hcl,bratton-marshall reagent,n-1-naphthalenyl-1,2-ethanediamine dihydrochloride
15106
CHEBI:53452
N'-naphthalen-1-ylethane-1,2-diamine;dihydrochloride
C1=CC=C2C(=C1)C=CC=C2NCCN.Cl.Cl
1465-25-4
C12H16Cl2N2
259.174
MZNYWPRCVDMOJG-UHFFFAOYSA-N
n-1-naphthyl ethylenediamine dihydrochloride,n1-naphthalen-1-yl ethane-1,2-diamine dihydrochloride,marshall's reagent,n-1-naphthylethylenediamine dihydrochloride,ccris 425,unii-h734599kjl,1,2-ethanediamine, n-1-naphthalenyl-, dihydrochloride,2-1-naphthylamino ethylamine 2hcl,bratton-marshall reagent,n-1-naphthalenyl-1,2-ethanediamine dihydrochloride
15106
CHEBI:53452
N'-naphthalen-1-ylethane-1,2-diamine;dihydrochloride
C1=CC=C2C(=C1)C=CC=C2NCCN.Cl.Cl

High quality chemicals for laboratory and specialized industrial use

    For Nitrogen Dioxide Determination (ASTM D-1607)
Packaging Wide Mouth Amber Glass Bottle
Quantity 100g
Formula Weight 259.18
Grade BAKER ANALYZED™ ACS Reagent
Chemical Name or Material N-1-Naphthylethylenediamine Dihydrochloride