Learn More
Medchemexpress LLC N-(1H-pyrrolo[2,3-c]pyridin-5-yl)benzamide | 300586-90-7 | MFCD00572806 | 99.9% | 237.26 | C14H11N3O | 50MG
SDP
Supplier:  Medchemexpress LLC HY1230350MG
Encompass_Preferred
OAC1 is a small-molecule Oct4 activator used in laboratory research to promote induced pluripotent stem cell (iPSC) formation by activating Oct4 and Nanog promoters. It is supplied as a solid and as a DMSO solution, and is intended for research use only.
  • Activates Oct4 and Nanog promoters.
  • Enhances induced pluripotent stem cell formation.
  • Available in solid and solution formats.
  • High purity suitable for biochemical assays.
  • Stable when stored under recommended conditions.
Catalog No. 50-002-35729