Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-[(1S)-2-Hydroxy-1-phenethyl)]ethoxycarboxamide, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
108493-65-8
C11H15NO3
209.245
MFCD06799380
GGXTXKPEZSYEOB-SNVBAGLBSA-N
ethyl n-1s-2-hydroxy-1-phenylethyl carbamate,s-2-ethoxycarbonylamino-2-phenylethanol,n-1s-2-hydroxy-1-phenethyl !ethoxycarboxamide,n-1s-2-hydroxy-1-phenethyl ethoxycarboxamide
8030353
ethyl N-[(1S)-2-hydroxy-1-phenylethyl]carbamate
CCOC(=O)NC(CO)C1=CC=CC=C1
108493-65-8
C11H15NO3
209.245
MFCD06799380
GGXTXKPEZSYEOB-SNVBAGLBSA-N
ethyl n-1s-2-hydroxy-1-phenylethyl carbamate,s-2-ethoxycarbonylamino-2-phenylethanol,n-1s-2-hydroxy-1-phenethyl !ethoxycarboxamide,n-1s-2-hydroxy-1-phenethyl ethoxycarboxamide
8030353
ethyl N-[(1S)-2-hydroxy-1-phenylethyl]carbamate
CCOC(=O)NC(CO)C1=CC=CC=C1
Quantity 1g
Packaging Glass bottle
Formula Weight 209.24
Chemical Name or Material N-[(1S)-2-Hydroxy-1-phenethyl)]ethoxycarboxamide

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.