Alfa Aesar™ N-(2,2,2-Trichloroethoxycarbonyloxy)succinimide, 98+%

Manufacturer: Alfa Aesar™ H5461606

Quantity 5g

 View more versions of this product

Catalog No. AAH5461606

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 66065-85-8
    Molecular Formula C7H6Cl3NO5
    Formula Weight 290.49
    MDL Number MFCD00075216
    Synonym N-Succinimidyl 2,2,2-trichloroethyl carbonate
    Chemical Name or Material N-(2,2,2-Trichloroethoxycarbonyloxy)succinimide

    Specifications

    Formula Weight 290.49
    Health Hazard 2 GHS H Statement
    H301-H311-H332-H319-H335
    Toxic if swallowed.
    Toxic in contact with skin.
    Harmful if inhaled.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Melting Point 111° to 113°C
    Quantity 5g
    Molecular Formula C7H6Cl3NO5
    MDL Number MFCD00075216
    CAS 66065-85-8
    Synonym N-Succinimidyl 2,2,2-trichloroethyl carbonate
    Health Hazard 1 Danger
    Health Hazard 3 GHS P Statement
    P261-P301+P310-P305+P351+P338-P361-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Recommended Storage 2° to 8°C
    Chemical Name or Material N-(2,2,2-Trichloroethoxycarbonyloxy)succinimide
    Assay ≥98%

    Description

    Applications
    N-(2,2,2-Trichloroethoxycarbonyloxy)succinimide was used in the synthesis of methyl [phenyl 5-(2,2,2-trichloroethoxycarbonylamino)-3,5-dideoxy-2-thio-β-D-glycero-D-galacto-2-nonulopyranosid]-onate; and phenyl 3,4-di-O-benzyl-6-O-(tert-butyl-dimethyl-silyl)-1-selanyl-2-(2,2,2-trichloroethoxycarbonylamine)-β-D-galactopyranoside.

    Solubility
    Soluble in dioxane, THF, DMF, CH2Cl2, alcohols, and most common organic solvents.

    Notes
    Moisture sensitive. Incompatible with oxidizing agents and heat.

    GHS H Statement
    H301-H311-H332-H319-H335
    Toxic if swallowed.
    Toxic in contact with skin.
    Harmful if inhaled.
    Causes serious eye irritation.
    May cause respiratory irritation.

    2° to 8°C

    GHS P Statement
    P261-P301+P310-P305+P351+P338-P361-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit