Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-(2,2,2-Trichloroethoxycarbonyloxy)succinimide, 98+%

Manufacturer:  Alfa Aesar™ H5461614

Quantity 25g

 View more versions of this product

Catalog No. AAH5461614

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Description

Applications
N-(2,2,2-Trichloroethoxycarbonyloxy)succinimide was used in the synthesis of methyl [phenyl 5-(2,2,2-trichloroethoxycarbonylamino)-3,5-dideoxy-2-thio-β-D-glycero-D-galacto-2-nonulopyranosid]-onate; and phenyl 3,4-di-O-benzyl-6-O-(tert-butyl-dimethyl-silyl)-1-selanyl-2-(2,2,2-trichloroethoxycarbonylamine)-β-D-galactopyranoside.

Solubility
Soluble in dioxane, THF, DMF, CH2Cl2, alcohols, and most common organic solvents.

Notes
Moisture sensitive. Incompatible with oxidizing agents and heat.
Description & Specifications

Chemical Identifiers

66065-85-8
290.49
N-Succinimidyl 2,2,2-trichloroethyl carbonate
C7H6Cl3NO5
MFCD00075216
N-(2,2,2-Trichloroethoxycarbonyloxy)succinimide

Specifications

111° to 113°C
66065-85-8
290.49
N-Succinimidyl 2,2,2-trichloroethyl carbonate
≥98%
25g
C7H6Cl3NO5
MFCD00075216
N-(2,2,2-Trichloroethoxycarbonyloxy)succinimide

Safety and Handling

GHS H Statement
H301-H311-H332-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

Recommended Storage : 2° to 8°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit