Alfa Aesar™ N-(2,4,6-Trimethylphenyl)thiourea, 99%

Manufacturer: Alfa Aesar™ A1208306

Quantity 5g

 View more versions of this product

Catalog No. AAA1208306

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 91059-97-1
    Molecular Formula C10H14N2S
    Formula Weight 194.3
    MDL Number MFCD00051488
    UN Number UN2811
    Beilstein 2642667
    Synonym N-Mesitylthiourea
    Chemical Name or Material N- (2,4,6-Trimethylphenyl)thiourea

    Specifications

    Formula Weight 194.3
    Health Hazard 3 GHS P Statement
    P280f-P301+P310-P330-P501a
    Wear protective clothing.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Rinse mouth.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H301
    Toxic if swallowed.
    MDL Number MFCD00051488
    UN Number UN2811
    Beilstein 2642667
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
    Melting Point 230° to 231°C
    Molecular Formula C10H14N2S
    CAS 91059-97-1
    Synonym N-Mesitylthiourea
    Health Hazard 1 Danger
    Quantity 5g
    Chemical Name or Material N- (2,4,6-Trimethylphenyl)thiourea
    Assay 99%

    GHS H Statement
    H301
    Toxic if swallowed.

    GHS P Statement
    P280f-P301+P310-P330-P501a
    Wear protective clothing.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Rinse mouth.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit