Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-(2,4,6-Trimethylphenyl)thiourea, 99%

Manufacturer:  Alfa Aesar™ A1208314

Quantity 25g

 View more versions of this product

Catalog No. AAA1208314


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

91059-97-1
194.3
UN2811
N-Mesitylthiourea
C10H14N2S
MFCD00051488
2642667
N- (2,4,6-Trimethylphenyl)thiourea

Specifications

230° to 231°C
91059-97-1
194.3
UN2811
N-Mesitylthiourea
99%
25g
C10H14N2S
MFCD00051488
2642667
N- (2,4,6-Trimethylphenyl)thiourea

Safety and Handling

SDS & Safety Documents

GHS H Statement
H301
Toxic if swallowed.

GHS P Statement
P280f-P301+P310-P330-P501a
Wear protective clothing.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit