Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-(2,4-Dichlorophenyl)maleamic acid, 97%

Click to view available options
Quantity:
1g
5g
25g
95695-47-9
C10H7Cl2NO3
260.07
MFCD00082905
WFWKZRKAECLVSE-ARJAWSKDSA-N
n-2,4-dichlorophenyl maleamic acid,2z-3-2,4-dichlorophenyl carbamoyl prop-2-enoic acid,4-2,4-dichloroanilino-4-oxobut-2-enoic acid,n-2,4-dichlorophenyl maleamidic acid,z-4-2,4-dichloroanilino-4-oxobut-2-enoic acid,z-4-2,4-dichlorophenyl amino-4-oxobut-2-enoic acid
1549984
(Z)-4-(2,4-dichloroanilino)-4-oxobut-2-enoic acid
C1=CC(=C(C=C1Cl)Cl)NC(=O)C=CC(=O)O
95695-47-9
C10H7Cl2NO3
260.07
MFCD00082905
WFWKZRKAECLVSE-ARJAWSKDSA-N
n-2,4-dichlorophenyl maleamic acid,2z-3-2,4-dichlorophenyl carbamoyl prop-2-enoic acid,4-2,4-dichloroanilino-4-oxobut-2-enoic acid,n-2,4-dichlorophenyl maleamidic acid,z-4-2,4-dichloroanilino-4-oxobut-2-enoic acid,z-4-2,4-dichlorophenyl amino-4-oxobut-2-enoic acid
1549984
(Z)-4-(2,4-dichloroanilino)-4-oxobut-2-enoic acid
C1=CC(=C(C=C1Cl)Cl)NC(=O)C=CC(=O)O
Quantity 1g
Melting Point 177°C to 180°C
Formula Weight 260.07
Percent Purity 97%
Chemical Name or Material N-(2,4-Dichlorophenyl)maleamic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures