Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-(2,4-Dichlorophenyl)maleamic acid, 97%

Click to view available options
Quantity:
1g
5g
25g
95695-47-9
C10H7Cl2NO3
260.07
MFCD00082905
WFWKZRKAECLVSE-ARJAWSKDSA-N
n-2,4-dichlorophenyl maleamic acid,2z-3-2,4-dichlorophenyl carbamoyl prop-2-enoic acid,4-2,4-dichloroanilino-4-oxobut-2-enoic acid,n-2,4-dichlorophenyl maleamidic acid,z-4-2,4-dichloroanilino-4-oxobut-2-enoic acid,z-4-2,4-dichlorophenyl amino-4-oxobut-2-enoic acid
1549984
(Z)-4-(2,4-dichloroanilino)-4-oxobut-2-enoic acid
C1=CC(=C(C=C1Cl)Cl)NC(=O)C=CC(=O)O
95695-47-9
C10H7Cl2NO3
260.07
MFCD00082905
WFWKZRKAECLVSE-ARJAWSKDSA-N
n-2,4-dichlorophenyl maleamic acid,2z-3-2,4-dichlorophenyl carbamoyl prop-2-enoic acid,4-2,4-dichloroanilino-4-oxobut-2-enoic acid,n-2,4-dichlorophenyl maleamidic acid,z-4-2,4-dichloroanilino-4-oxobut-2-enoic acid,z-4-2,4-dichlorophenyl amino-4-oxobut-2-enoic acid
1549984
(Z)-4-(2,4-dichloroanilino)-4-oxobut-2-enoic acid
C1=CC(=C(C=C1Cl)Cl)NC(=O)C=CC(=O)O
Melting Point 177°C to 180°C
Quantity 25g
Formula Weight 260.07
Percent Purity 97%
Chemical Name or Material N-(2,4-Dichlorophenyl)maleamic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning