Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-(2,4-Difluorophenyl)thiourea, 97%

Click to view available options
Quantity:
1g
5g
175277-76-6
C7H6F2N2S
188.196
MFCD00041150
DZZSKQFBAGZNSH-UHFFFAOYSA-N
2,4-difluorophenyl thiourea,1-2,4-difluorophenyl thiourea,1-2,4-difluorophenyl-2-thiourea,n-2,4-difluorophenyl thiourea,thiourea,n-2,4-difluorophenyl,amino 2,4-difluorophenyl amino methane-1-thione,acmc-1bxvl,buttpark 12118-31,difluorophenylthioacetonitrile,2,4-difluoro-phenyl-thiourea
2734207
(2,4-difluorophenyl)thiourea
C1=CC(=C(C=C1F)F)NC(=S)N
175277-76-6
C7H6F2N2S
188.196
MFCD00041150
DZZSKQFBAGZNSH-UHFFFAOYSA-N
2,4-difluorophenyl thiourea,1-2,4-difluorophenyl thiourea,1-2,4-difluorophenyl-2-thiourea,n-2,4-difluorophenyl thiourea,thiourea,n-2,4-difluorophenyl,amino 2,4-difluorophenyl amino methane-1-thione,acmc-1bxvl,buttpark 12118-31,difluorophenylthioacetonitrile,2,4-difluoro-phenyl-thiourea
2734207
(2,4-difluorophenyl)thiourea
C1=CC(=C(C=C1F)F)NC(=S)N
Melting Point 165°C to 168°C
Quantity 1g
UN Number UN2811
Formula Weight 188.2
Percent Purity 97%
Chemical Name or Material N-(2,4-Difluorophenyl)thiourea

GHS H Statement
H301
Toxic if swallowed.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c

H302+H312+H332

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures