Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-(2,6-Dichlorophenyl)thiourea, 99%

Click to view available options
Quantity:
5g
25g
6590-91-6
C7H6Cl2N2S
221.099
MFCD00041156
KUQHRGMPBWZVQR-UHFFFAOYSA-N
1-2,6-dichlorophenyl thiourea,2,6-dichlorophenyl thiourea,n-2,6-dichlorophenyl thiourea,thiourea, 2,6-dichlorophenyl,1-2,6-dichlorophenyl-2-thiourea,amino 2,6-dichlorophenyl amino methane-1-thione,chembl29234,2,6-dichlorophenyl-thiourea,thiourea,n-2,6-dichlorophenyl
722776
(2,6-dichlorophenyl)thiourea
C1=CC(=C(C(=C1)Cl)NC(=S)N)Cl
6590-91-6
C7H6Cl2N2S
221.099
MFCD00041156
KUQHRGMPBWZVQR-UHFFFAOYSA-N
1-2,6-dichlorophenyl thiourea,2,6-dichlorophenyl thiourea,n-2,6-dichlorophenyl thiourea,thiourea, 2,6-dichlorophenyl,1-2,6-dichlorophenyl-2-thiourea,amino 2,6-dichlorophenyl amino methane-1-thione,chembl29234,2,6-dichlorophenyl-thiourea,thiourea,n-2,6-dichlorophenyl
722776
(2,6-dichlorophenyl)thiourea
C1=CC(=C(C(=C1)Cl)NC(=S)N)Cl
Melting Point 155°C to 159°C
Quantity 5g
Odor Odorless
UN Number UN2811
Beilstein 2212144
Formula Weight 221.11
Percent Purity 99%
Chemical Name or Material N-(2,6-Dichlorophenyl)thiourea

GHS H Statement
H301
Toxic if swallowed.

P264b-P270-P301+P310-P330-P501c

H301

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures