Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(2-Aminoethyl)aminomethyl Polystyrene Resin cross-linked with 1% DVB (50-100mesh) (3.1-3.5mmol/g), TCI America™

Catalog Number A200625G
Click to view available options
Quantity:
5 g
25 g
MFCD01635304
MFCD01635304
Color Yellow
Quantity 25 g
Physical Form Crystalline Powder
Chemical Name or Material N-(2-Aminoethyl)aminomethyl Polystyrene Resin cross-linked with 1% DVB (50-100mesh) (3.1-3.5mmol/g)
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit