Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(2-Aminoethyl)aminomethyl Polystyrene Resin cross-linked with 1% DVB (50-100mesh) (3.1-3.5mmol/g), TCI America™

Click to view available options
Quantity:
5g
25g
MFCD01635304
MFCD01635304
Color Yellow
Quantity 5g
Physical Form Crystal-Powder at 20°C
Chemical Name or Material N-(2-Aminoethyl)aminomethyl Polystyrene Resin cross-linked with 1% DVB (50-100mesh) (3.1-3.5mmol/g)