Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(2-Chloroethyl)acetamide, 98%, ACROS Organics™

Click to view available options
Quantity:
10g
50g
7355-58-0
C4H8ClNO
121.57
MFCD00000965
HSKNJSHFPPHTAQ-UHFFFAOYSA-N
n-2-chloroethyl acetamide,acetamide, n-2-chloroethyl,n-2-chloro-ethyl-acetamide,n-acetyl-2-chloroethylamine,n 2-chloroethyl acetamide,n-acetyl-ss-chlorathylamin,acmc-1bi05,n-2-chloroethyl-acetamide,4-04-00-00449 beilstein handbook reference
81815
N-(2-chloroethyl)acetamide
CC(=O)NCCCl
7355-58-0
C4H8ClNO
121.57
MFCD00000965
HSKNJSHFPPHTAQ-UHFFFAOYSA-N
n-2-chloroethyl acetamide,acetamide, n-2-chloroethyl,n-2-chloro-ethyl-acetamide,n-acetyl-2-chloroethylamine,n 2-chloroethyl acetamide,n-acetyl-ss-chlorathylamin,acmc-1bi05,n-2-chloroethyl-acetamide,4-04-00-00449 beilstein handbook reference
81815
N-(2-chloroethyl)acetamide
CC(=O)NCCCl
Specific Gravity 1.2
Infrared Spectrum Authentic
Quantity 10g
Flash Point 110°C
Density 1.2000g/mL
Refractive Index 1.4770 to 1.48
Color Colorless to Yellow
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Linear Formula CH3CONHCH2CH2Cl
Formula Weight 121.57
Physical Form Viscous Liquid
Percent Purity 98%
Chemical Name or Material N-(2-Chloroethyl)acetamide, 98%
Show More Show Less

GHS H Statement:
May cause damage to organs through prolonged or repeated exposure.

GHS P Statement:
Do not breathe dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 230-884-5

RTECSNumber : AB5460000