Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(2-Chlorophenyl)maleimide, 97%, Acros Organics™

Click to view available options
Quantity:
1g
5g
1203-24-3
C10H6ClNO2
207.62
KPQOXMCRYWDRSB-UHFFFAOYSA-N
1-2-chlorophenyl-1h-pyrrole-2,5-dione,n-2-chlorophenyl maleimide,o-chphm,o-chlorophenylmaleimide,n-o-chlorophenylmaleimide,1-2-chlorophenyl pyrrole-2,5-dione,1h-pyrrole-2,5-dione, 1-2-chlorophenyl,n-2-chlorophenyl-maleimide,maleimide, n-o-chlorophenyl,n-o-chlorophenyl maleiimide
14550
1-(2-chlorophenyl)pyrrole-2,5-dione
C1=CC=C(C(=C1)N2C(=O)C=CC2=O)Cl
1203-24-3
C10H6ClNO2
207.62
KPQOXMCRYWDRSB-UHFFFAOYSA-N
1-2-chlorophenyl-1h-pyrrole-2,5-dione,n-2-chlorophenyl maleimide,o-chphm,o-chlorophenylmaleimide,n-o-chlorophenylmaleimide,1-2-chlorophenyl pyrrole-2,5-dione,1h-pyrrole-2,5-dione, 1-2-chlorophenyl,n-2-chlorophenyl-maleimide,maleimide, n-o-chlorophenyl,n-o-chlorophenyl maleiimide
14550
1-(2-chlorophenyl)pyrrole-2,5-dione
C1=CC=C(C(=C1)N2C(=O)C=CC2=O)Cl
Infrared Spectrum Authentic
Quantity 5g
Melting Point 108.0°C to 109.5°C
Assay Percent Range 96% min. (GC)
Formula Weight 207.62
Percent Purity 97%
Chemical Name or Material N-(2-Chlorophenyl)maleimide

GHS H Statement Harmful if swallowed. Harmful in contact with skin. Causes skin irritation. Causes serious eye irritation. Harmful if inhaled. May cause respiratory irritation.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.

GHS Signal Word: Warning

EINECSNumber : 214-869-