Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-(2-Hydroxy-4-methylphenyl)acetamide, 97%

Click to view available options
Quantity:
250mg
1g
13429-10-2
C9H11NO2
165.192
MFCD00012061
HTVPAACTHAQQAS-UHFFFAOYSA-N
n-2-hydroxy-4-methylphenyl acetamide,acetamide, n-2-hydroxy-4-methylphenyl,acetamide,n-2-hydroxy-4-methylphenyl,6-acetamino-m-kresol,2'-hydroxy-p-acetotoluidide,2'-hydroxy-4'-methylacetoanilide,2-acetamido-5-methylphenol,n-2-hydroxy-4-methyl-phenyl-acetamide
83437
N-(2-hydroxy-4-methylphenyl)acetamide
CC1=CC(=C(C=C1)NC(=O)C)O
13429-10-2
C9H11NO2
165.192
MFCD00012061
HTVPAACTHAQQAS-UHFFFAOYSA-N
n-2-hydroxy-4-methylphenyl acetamide,acetamide, n-2-hydroxy-4-methylphenyl,acetamide,n-2-hydroxy-4-methylphenyl,6-acetamino-m-kresol,2'-hydroxy-p-acetotoluidide,2'-hydroxy-4'-methylacetoanilide,2-acetamido-5-methylphenol,n-2-hydroxy-4-methyl-phenyl-acetamide
83437
N-(2-hydroxy-4-methylphenyl)acetamide
CC1=CC(=C(C=C1)NC(=O)C)O
Quantity 1g
Beilstein 972595
Formula Weight 165.19
Physical Form Solid
Percent Purity 97%
Chemical Name or Material N-(2-Hydroxy-4-methylphenyl)acetamide

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA