Alfa Aesar™ N-(2-Hydroxybenzoyl)pyrrolidine, 97%

Manufacturer: Alfa Aesar™ H5569514

Quantity 25g

 View more versions of this product

Catalog No. AAH5569514

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 98841-68-0
    Formula Weight 191.23
    MDL Number MFCD00020832
    Chemical Name or Material N-(2-Hydroxybenzoyl)pyrrolidine

    Specifications

    Melting Point 118° to 124°C
    Formula Weight 191.23
    Health Hazard 3 GHS P Statement
    P280-P264-P305+P351+P338-P301+P312-P337+P313-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash thoroughly after handling.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    If eye irritation persists: Get medical advice/attention.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Solid
    MDL Number MFCD00020832
    Health Hazard 1 Warning
    Color White to pale brown
    CAS 98841-68-0
    Health Hazard 2 GHS H Statement
    H302-H319
    Harmful if swallowed.
    Causes serious eye irritation.
    Quantity 25g
    Chemical Name or Material N-(2-Hydroxybenzoyl)pyrrolidine
    Assay 97%

    GHS H Statement
    H302-H319
    Harmful if swallowed.
    Causes serious eye irritation.

    GHS P Statement
    P280-P264-P305+P351+P338-P301+P312-P337+P313-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash thoroughly after handling.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    If eye irritation persists: Get medical advice/attention.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit