Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(2-Hydroxyethyl)ethylenediaminetriacetic acid, trisodium salt hydrate, 85%, ACROS Organics™

Click to view available options
Quantity:
5g
207386-87-6
C10H17N2Na3O8
362.221
MFCD00150694
FHGJYTNPLCNRLG-UHFFFAOYSA-K
trisodium hedta monohydrate,unii-463a7rib6k,n-2-hydroxyethyl ethylenediaminetriacetic acid trisodium salt hydrate,glycine, n-2-bis carboxymethyl amino ethyl-n-2-hydroxyethyl-, trisodium salt hydrate,trisodium 2-2-bis carboxylatomethyl amino ethyl 2-hydroxyethyl amino acetate hydrate,sodium 2,2'-2-carboxylatomethyl 2-hydroxyethyl amino ethylazanediyl diacetate hydrate,glycine, n-2-bis carboxymethyl amino ethyl-n-2-hydroxyethyl-, trisodium salt, monohydrate
16211347
trisodium;2-[2-[bis(carboxylatomethyl)amino]ethyl-(2-hydroxyethyl)amino]acetate;hydrate
C(CN(CC(=O)[O-])CC(=O)[O-])N(CCO)CC(=O)[O-].O.[Na+].[Na+].[Na+]
207386-87-6
C10H17N2Na3O8
362.221
MFCD00150694
FHGJYTNPLCNRLG-UHFFFAOYSA-K
trisodium hedta monohydrate,unii-463a7rib6k,n-2-hydroxyethyl ethylenediaminetriacetic acid trisodium salt hydrate,glycine, n-2-bis carboxymethyl amino ethyl-n-2-hydroxyethyl-, trisodium salt hydrate,trisodium 2-2-bis carboxylatomethyl amino ethyl 2-hydroxyethyl amino acetate hydrate,sodium 2,2'-2-carboxylatomethyl 2-hydroxyethyl amino ethylazanediyl diacetate hydrate,glycine, n-2-bis carboxymethyl amino ethyl-n-2-hydroxyethyl-, trisodium salt, monohydrate
16211347
trisodium;2-[2-[bis(carboxylatomethyl)amino]ethyl-(2-hydroxyethyl)amino]acetate;hydrate
C(CN(CC(=O)[O-])CC(=O)[O-])N(CCO)CC(=O)[O-].O.[Na+].[Na+].[Na+]
Infrared Spectrum Authentic
Quantity 5g
Color White to Yellow
CAS Min % 80.0
CAS Max % 100.0
Assay Percent Range 85%
Merck Index 14, 9966
Formula Weight 344.21
Physical Form Crystalline Powder
Percent Purity ≥80%
Chemical Name or Material N-(2-Hydroxyethyl)ethylenediaminetriacetic acid, trisodium salt hydrate
Show More Show Less

GHS H Statement:
Causes serious eye damage.
Harmful if swallowed.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 205-381-9

RTECSNumber : MB9188000

TSCA : TSCA