Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(2-Hydroxyethyl)phthalimide, 99%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
3891-07-4
C10H9NO3
191.19
MFCD00005903
MWFLUYFYHANMCM-UHFFFAOYSA-N
2-1,2,2-trimethylpropylcarbamoyl benzoic acid,2-3,3-dimethylbutan-2-yl carbamoyl benzoic acid,2-1,2,2-trimethylpropyl amino carbonyl benzoic acid,n-pinacolylphthalamic acid,2-3,3-dimethylbutan-2-ylcarbamoyl benzoic acid,+-n-pinacolylphthalamic acid
3354762
2-(3,3-dimethylbutan-2-ylcarbamoyl)benzoic acid
OCCN1C(=O)C2=CC=CC=C2C1=O
3891-07-4
C10H9NO3
191.19
MFCD00005903
MWFLUYFYHANMCM-UHFFFAOYSA-N
2-1,2,2-trimethylpropylcarbamoyl benzoic acid,2-3,3-dimethylbutan-2-yl carbamoyl benzoic acid,2-1,2,2-trimethylpropyl amino carbonyl benzoic acid,n-pinacolylphthalamic acid,2-3,3-dimethylbutan-2-ylcarbamoyl benzoic acid,+-n-pinacolylphthalamic acid
3354762
2-(3,3-dimethylbutan-2-ylcarbamoyl)benzoic acid
OCCN1C(=O)C2=CC=CC=C2C1=O
Infrared Spectrum Authentic
Quantity 25g
Packaging Glass bottle
Melting Point 126°C to 129°C
Color White
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (on dry substance) (HPLC)
Formula Weight 191.19
Physical Form Crystalline Powder
Percent Purity 99%
Chemical Name or Material N-(2-Hydroxyethyl)phthalimide
Show More Show Less

EINECSNumber : 223-434-4