Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-[2-(Piperidinylamino)ethyl]-4-iodobenzamide, Tocris Bioscience™

Click to view available options
Quantity:
10mg
50mg

Chemical Identifiers

155054-42-5
C14H19IN2O
358.223
WCMWVYQHPUQKHW-UHFFFAOYSA-N
127898
4-iodo-N-(2-piperidin-1-ylethyl)benzamide
C1CCN(CC1)CCNC(=O)C2=CC=C(C=C2)I

σ1 selective ligand

σ-1 selective ligand which binds with high affinity to human malignant melanoma cells A 2058.
Quantity 10mg
Formula Weight 358.22
Chemical Name or Material N-[2-(Piperidinylamino)ethyl]-4-iodobenzamide
Grade >98%

Recommended Storage : Desiccate at +4°C