Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(2-Pyridyl)bis(trifluoromethanesulfonimide), 97%, Acros Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
145100-50-1
C7H4F6N2O4S2
358.24
MFCD00191834
DXLQEJHUQKKSRB-UHFFFAOYSA-N
2-n,n-bis trifluoromethylsulfonyl amino pyridine,1,1,1-trifluoro-n-pyridin-2-yl-n-trifluoromethyl sulfonyl methanesulfonamide,n-pyridin-2-yl triflimide,2-n,n-bis trifluoromethyl sulphonylamino pyridine,n-2-pyridyl triflimide,n-2-pyridyl bis trifluoromethanesulfonimide,pyridin-2-amine, n,n-bis trifluoromethylsulfonyl,2-bis trifluoromethylsulfonyl amino pyridine,1,1,1-trifluoro-n-pyridin-2-yl-n-trifluoromethanesulfonylmethanesulfonamide,n,n-bis trifluoromethylsulfonyl-2-pyridylamine
534172
1,1,1-trifluoro-N-pyridin-2-yl-N-(trifluoromethylsulfonyl)methanesulfonamide
C1=CC=NC(=C1)N(S(=O)(=O)C(F)(F)F)S(=O)(=O)C(F)(F)F
145100-50-1
C7H4F6N2O4S2
358.24
MFCD00191834
DXLQEJHUQKKSRB-UHFFFAOYSA-N
2-n,n-bis trifluoromethylsulfonyl amino pyridine,1,1,1-trifluoro-n-pyridin-2-yl-n-trifluoromethyl sulfonyl methanesulfonamide,n-pyridin-2-yl triflimide,2-n,n-bis trifluoromethyl sulphonylamino pyridine,n-2-pyridyl triflimide,n-2-pyridyl bis trifluoromethanesulfonimide,pyridin-2-amine, n,n-bis trifluoromethylsulfonyl,2-bis trifluoromethylsulfonyl amino pyridine,1,1,1-trifluoro-n-pyridin-2-yl-n-trifluoromethanesulfonylmethanesulfonamide,n,n-bis trifluoromethylsulfonyl-2-pyridylamine
534172
1,1,1-trifluoro-N-pyridin-2-yl-N-(trifluoromethylsulfonyl)methanesulfonamide
C1=CC=NC(=C1)N(S(=O)(=O)C(F)(F)F)S(=O)(=O)C(F)(F)F
Coupling reagent
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (HPLC)
Fieser 17,293
Formula Weight 358.24
Physical Form Crystalline Powder or Crystals
Percent Purity 97%
Density 110g/mL
Infrared Spectrum Authentic
Packaging Glass bottle
Boiling Point 97.0°C (2.0 mmHg)
Color White
Melting Point 40.0°C to 43.0°C
Quantity 1g
Flash Point 110°C
Chemical Name or Material N-(2-Pyridyl)bis(trifluoromethanesulfonimide)
Show More Show Less

GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning