Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(3-Aminopropyl)cyclohexylamine 98.0+%, TCI America™

Click to view available options
Quantity:
25mL
500mL
3312-60-5
C9H20N2
156.273
MFCD00003838
ITZPOSYADVYECJ-UHFFFAOYSA-N
n-3-aminopropyl cyclohexylamine,n1-cyclohexylpropane-1,3-diamine,1,3-propanediamine, n-cyclohexyl,n-cyclohexyl-1,3-propanediamine,cyclohexyl-1,3-propanediamine,3-aminopropyl cyclohexylamine,n-cyclohexyl-1,3-diaminopropane,unii-f4xob09fkn,n-cyclohexyl-1,3-propylenediamine,n-cyclohexylpropylene 1,3-diamine
18718
N'-cyclohexylpropane-1,3-diamine
C1CCC(CC1)NCCCN
3312-60-5
C9H20N2
156.273
MFCD00003838
ITZPOSYADVYECJ-UHFFFAOYSA-N
n-3-aminopropyl cyclohexylamine,n1-cyclohexylpropane-1,3-diamine,1,3-propanediamine, n-cyclohexyl,n-cyclohexyl-1,3-propanediamine,cyclohexyl-1,3-propanediamine,3-aminopropyl cyclohexylamine,n-cyclohexyl-1,3-diaminopropane,unii-f4xob09fkn,n-cyclohexyl-1,3-propylenediamine,n-cyclohexylpropylene 1,3-diamine
18718
N'-cyclohexylpropane-1,3-diamine
C1CCC(CC1)NCCCN
Quantity 25mL
Boiling Point 123°C
Color Yellow
UN Number 2735
Formula Weight 156.27
Physical Form Clear Liquid at 20°C
Percent Purity ≥98.0% (GC,T)
Chemical Name or Material N-(3-Aminopropyl)cyclohexylamine