Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(3-Aminopropyl)glycine Dihydrochloride 97.0+%, TCI America™

Click to view available options
Quantity:
1g
90495-95-7
C5H14Cl2N2O2
205.079
MFCD00144831
JQQTXSWVGCRXGJ-UHFFFAOYSA-N
N-(3-Aminopropyl)aminoacetic Acid Dihydrochloride
44630059
2-(3-aminopropylamino)acetic acid;dihydrochloride
C(CN)CNCC(=O)O.Cl.Cl
90495-95-7
C5H14Cl2N2O2
205.079
MFCD00144831
JQQTXSWVGCRXGJ-UHFFFAOYSA-N
N-(3-Aminopropyl)aminoacetic Acid Dihydrochloride
44630059
2-(3-aminopropylamino)acetic acid;dihydrochloride
C(CN)CNCC(=O)O.Cl.Cl
Quantity 1g
Color White-Yellow
Formula Weight 205.08
Physical Form Crystal-Powder at 20°C
Percent Purity ≥97.0% (T)
Chemical Name or Material N-(3-Aminopropyl)glycine Dihydrochloride