Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(3-Aminopropyl)pyrrolidine, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
23159-07-1
C7H16N2
128.22
MFCD00014100
VPBWZBGZWHDNKL-UHFFFAOYSA-N
3-pyrrolidin-1-yl propan-1-amine,1-3-aminopropyl pyrrolidine,1-3-aminopropyl-pyrrolidine,1-pyrrolidinepropanamine,n-3-aminopropyl pyrrolidine,3-pyrrolidinopropylamine,3-1-pyrrolidinyl propylamine,pyrrolidine, 1-3-aminopropyl,3-pyrrolidin-1-ylpropylamine,3-1-pyrrolidino propylamine
31670
3-pyrrolidin-1-ylpropan-1-amine
C1CCN(C1)CCCN
23159-07-1
C7H16N2
128.22
MFCD00014100
VPBWZBGZWHDNKL-UHFFFAOYSA-N
3-pyrrolidin-1-yl propan-1-amine,1-3-aminopropyl pyrrolidine,1-3-aminopropyl-pyrrolidine,1-pyrrolidinepropanamine,n-3-aminopropyl pyrrolidine,3-pyrrolidinopropylamine,3-1-pyrrolidinyl propylamine,pyrrolidine, 1-3-aminopropyl,3-pyrrolidin-1-ylpropylamine,3-1-pyrrolidino propylamine
31670
3-pyrrolidin-1-ylpropan-1-amine
C1CCN(C1)CCCN
Infrared Spectrum Authentic
Boiling Point 191°C to 193°C
Specific Gravity 1.014
Refractive Index 1.4700 to 1.472
Density 1.0140g/mL
Quantity 5g
Color Yellow
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Solubility Information Solubility in water: soluble. Other solubilities: soluble in chloroform
Formula Weight 128.22
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material N-(3-Aminopropyl)pyrrolidine
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 245-465-2