Alfa Aesar™ N-(3-Chlorophenylsulfonyl)-DL-phenylalanine, 96%

Manufacturer: Alfa Aesar™ H33999MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH33999MD

  Check Availability
Add to Cart

    Chemical Identifiers

    Molecular Formula C15H14ClNO4S
    Formula Weight 339.8
    MDL Number MFCD07433680
    Chemical Name or Material N-(3-Chlorophenylsulfonyl)-DL-phenylalanine

    Specifications

    Formula Weight 339.8
    Quantity 250mg
    Molecular Formula C15H14ClNO4S
    MDL Number MFCD07433680
    Melting Point 121° to 123°C
    Chemical Name or Material N-(3-Chlorophenylsulfonyl)-DL-phenylalanine
    Assay 96%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit