Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-[3-(Trimethoxysilyl)propyl]-N,N,N-trimethylammonium chloride, in 50% methanol, Alfa Aesar™

Click to view available options
Quantity:
25g
100g
500g
35141-36-7
C9H24ClNO3Si
257.83
MFCD00054225
FYZFRYWTMMVDLR-UHFFFAOYSA-M
n-trimethoxysilylpropyl-n,n,n-trimethylammonium chloride,n,n,n-trimethyl-3-trimethoxysilyl propan-1-aminium chloride,1-propanaminium, n,n,n-trimethyl-3-trimethoxysilyl-, chloride,trimethyl-3-trimethoxysilyl propylammonium chloride,trimethyl 3-trimethoxysilyl propyl azanium chloride,1-propanaminium,n,n,n-trimethyl-3-trimethoxysilyl-, chloride 1:1,1-propanaminium, n,n,n-trimethyl-3-trimethoxysilyl-, chloride 1:1,3-trimethoxysilylpropyl trimethylammonium chloride,acmc-20ak0r,trimethyl-3-trimethoxysilyl propyl ammonium
94462
trimethyl(3-trimethoxysilylpropyl)azanium;chloride
C[N+](C)(C)CCC[Si](OC)(OC)OC.[Cl-]
35141-36-7
C9H24ClNO3Si
257.83
MFCD00054225
FYZFRYWTMMVDLR-UHFFFAOYSA-M
n-trimethoxysilylpropyl-n,n,n-trimethylammonium chloride,n,n,n-trimethyl-3-trimethoxysilyl propan-1-aminium chloride,1-propanaminium, n,n,n-trimethyl-3-trimethoxysilyl-, chloride,trimethyl-3-trimethoxysilyl propylammonium chloride,trimethyl 3-trimethoxysilyl propyl azanium chloride,1-propanaminium,n,n,n-trimethyl-3-trimethoxysilyl-, chloride 1:1,1-propanaminium, n,n,n-trimethyl-3-trimethoxysilyl-, chloride 1:1,3-trimethoxysilylpropyl trimethylammonium chloride,acmc-20ak0r,trimethyl-3-trimethoxysilyl propyl ammonium
94462
trimethyl(3-trimethoxysilylpropyl)azanium;chloride
C[N+](C)(C)CCC[Si](OC)(OC)OC.[Cl-]
Quantity 500g
Density 0.930g/mL
Color Colorless to Yellow
Refractive Index 1.3970
Sensitivity Hygroscopic
UN Number UN1230
Beilstein 11593127
Formula Weight 257.83
Physical Form Liquid
Chemical Name or Material N-[3-(Trimethoxysilyl)propyl]-N,N,N-trimethylammonium chloride, in 50% methanol

GHS H Statement
H225-H311-H331-H370-H335-H302-H315-H319
Highly flammable liquid and vapor.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P210-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packaging Group: II; Proper Shipping Name: METHANOL

TSCA : TSCA