Alfa Aesar™ N-(4-Aminobenzoyl)-L-glutamic acid diethyl ester, 99%

Manufacturer: Alfa Aesar™ B22305-09

Quantity 10g

 View more versions of this product

Catalog No. AAB2230509

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13726-52-8
    Molecular Formula C16H22N2O5
    Formula Weight 322.36g/mol
    MDL Number MFCD00009617
    Synonym Diethyl N-(4-aminobenzoyl)-L-glutamate
    Chemical Name or Material N-(4-Aminobenzoyl)-L-glutamic acid diethyl ester

    Specifications

    Melting Point 139° to 142°C
    Quantity 10g
    Optical Rotation 19° (c=1 Chloroform)
    CAS 13726-52-8
    Molecular Formula C16H22N2O5
    Formula Weight 322.36g/mol
    MDL Number MFCD00009617
    Synonym Diethyl N-(4-aminobenzoyl)-L-glutamate
    Chemical Name or Material N-(4-Aminobenzoyl)-L-glutamic acid diethyl ester
    Assay 99%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit