Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-(4-Aminobenzoyl)-L-glutamic acid diethyl ester, 99%

Manufacturer:  Alfa Aesar™ B2230530

Quantity 250g

 View more versions of this product

Catalog No. AAB2230530


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

13726-52-8
322.36
Diethyl N-(4-aminobenzoyl)-L-glutamate
C16H22N2O5
MFCD00009617
N-(4-Aminobenzoyl)-L-glutamic acid diethyl ester

Specifications

13726-52-8
322.36
Diethyl N-(4-aminobenzoyl)-L-glutamate
250g
N-(4-Aminobenzoyl)-L-glutamic acid diethyl ester
C16H22N2O5
MFCD00009617
139° to 142°C
19° (c=1 Chloroform)
99%
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit