Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-(4-Aminobenzoyl)-L-glutamic acid diethyl ester, 99%

Click to view available options
Quantity:
10g
50g
250g
13726-52-8
C16H22N2O5
322.361
MFCD00009617
RJXFBLRRPYBPTM-ZDUSSCGKSA-N
n-4-aminobenzoyl-l-glutamic acid diethyl ester,diethyl n-4-aminobenzoyl-l-glutamate,diethyl n-p-aminobenzoyl-l-glutamate,1,5-diethyl 2s-2-4-aminophenyl formamido pentanedioate,4-abz-glu oet-oet,l-glutamic acid, n-4-aminobenzoyl-, diethyl ester,diethyl 4-aminobenzoyl-l-glutamate,diethyl p-aminobenzoyl-l-glutamate,s-diethyl 2-4-aminobenzamido pentanedioate,diethyl 2-4-aminobenzoyl amino pentanedioate #
73002
diethyl (2S)-2-[(4-aminobenzoyl)amino]pentanedioate
CCOC(=O)CCC(C(=O)OCC)NC(=O)C1=CC=C(C=C1)N
13726-52-8
C16H22N2O5
322.361
MFCD00009617
RJXFBLRRPYBPTM-ZDUSSCGKSA-N
n-4-aminobenzoyl-l-glutamic acid diethyl ester,diethyl n-4-aminobenzoyl-l-glutamate,diethyl n-p-aminobenzoyl-l-glutamate,1,5-diethyl 2s-2-4-aminophenyl formamido pentanedioate,4-abz-glu oet-oet,l-glutamic acid, n-4-aminobenzoyl-, diethyl ester,diethyl 4-aminobenzoyl-l-glutamate,diethyl p-aminobenzoyl-l-glutamate,s-diethyl 2-4-aminobenzamido pentanedioate,diethyl 2-4-aminobenzoyl amino pentanedioate #
73002
diethyl (2S)-2-[(4-aminobenzoyl)amino]pentanedioate
CCOC(=O)CCC(C(=O)OCC)NC(=O)C1=CC=C(C=C1)N
Formula Weight 322.36
Percent Purity 99%
Melting Point 139°C to 142°C
Quantity 50g
Optical Rotation 19° (c=1 Chloroform)
Chemical Name or Material N-(4-Aminobenzoyl)-L-glutamic acid diethyl ester