Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-(4-Bromophenyl)maleamic acid, 97%

Click to view available options
Quantity:
1g
5g
25g
36847-86-6
C10H8BrNO3
270.082
MFCD00134952
DSNFUZCLEZJCRN-WAYWQWQTSA-N
n-4-bromophenyl maleamic acid,4-4-bromoanilino-4-oxobut-2-enoic acid,4-4-bromophenyl amino-4-oxobut-2-enoic acid,2z-3-4-bromophenyl carbamoyl prop-2-enoic acid,3-4-bromophenyl carbamoyl prop-2-enoic acid,n-4-bromophenyl maleamidic acid,3-4-bromo-phenylcarbamoyl-acrylic acid,z-4-4-bromoanilino-4-oxobut-2-enoic acid,2-butenoic acid, 4-4-bromophenyl amino-4-oxo-, 2z
1551067
(Z)-4-(4-bromoanilino)-4-oxobut-2-enoic acid
C1=CC(=CC=C1NC(=O)C=CC(=O)O)Br
36847-86-6
C10H8BrNO3
270.082
MFCD00134952
DSNFUZCLEZJCRN-WAYWQWQTSA-N
n-4-bromophenyl maleamic acid,4-4-bromoanilino-4-oxobut-2-enoic acid,4-4-bromophenyl amino-4-oxobut-2-enoic acid,2z-3-4-bromophenyl carbamoyl prop-2-enoic acid,3-4-bromophenyl carbamoyl prop-2-enoic acid,n-4-bromophenyl maleamidic acid,3-4-bromo-phenylcarbamoyl-acrylic acid,z-4-4-bromoanilino-4-oxobut-2-enoic acid,2-butenoic acid, 4-4-bromophenyl amino-4-oxo-, 2z
1551067
(Z)-4-(4-bromoanilino)-4-oxobut-2-enoic acid
C1=CC(=CC=C1NC(=O)C=CC(=O)O)Br
Quantity 25g
Formula Weight 270.08
Percent Purity 97%
Chemical Name or Material N-(4-Bromophenyl)maleamic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures