Alfa Aesar™ N-(4-Ethoxyphenyl)maleamic acid, 97%

Manufacturer: Alfa Aesar™ B2502406

Quantity 5g

 View more versions of this product

Catalog No. AAB2502406

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 108087-84-9
    Molecular Formula C12H13NO4
    Formula Weight 235.24
    MDL Number MFCD00020477
    Synonym 4-(4-Ethoxyanilino)-4-oxobut-2-enoic acid
    Chemical Name or Material N-(4-Ethoxyphenyl)maleamic acid

    Specifications

    Formula Weight 235.24
    CAS 108087-84-9
    Quantity 5g
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula C12H13NO4
    Melting Point 188° to 189°C
    MDL Number MFCD00020477
    Synonym 4-(4-Ethoxyanilino)-4-oxobut-2-enoic acid
    Health Hazard 1 Warning
    Chemical Name or Material N-(4-Ethoxyphenyl)maleamic acid
    Assay 97%

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit