Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-(4-Ethoxyphenyl)maleamic acid, 97%

Click to view available options
Quantity:
5g
25g
100g
108087-84-9
C12H13NO4
235.239
MFCD00020477
LMTIUWRMRAZPEN-FPLPWBNLSA-N
z-4-4-ethoxyphenyl amino-4-oxobut-2-enoic acid,2z-4-4-ethoxyphenyl amino-4-oxobut-2-enoic acid,n-4-ethoxyphenyl maleamic acid,2z-3-n-4-ethoxyphenyl carbamoyl prop-2-enoic acid,2z-3-4-ethoxyphenyl carbamoyl prop-2-enoic acid,4-4-ethoxyanilino-4-oxobut-2-enoic acid,z-4-4-ethoxyanilino-4-oxobut-2-enoic acid,2-butenoic acid,4-4-ethoxyphenyl amino-4-oxo-, 2z
1808883
(Z)-4-(4-ethoxyanilino)-4-oxobut-2-enoic acid
CCOC1=CC=C(C=C1)NC(=O)C=CC(=O)O
108087-84-9
C12H13NO4
235.239
MFCD00020477
LMTIUWRMRAZPEN-FPLPWBNLSA-N
z-4-4-ethoxyphenyl amino-4-oxobut-2-enoic acid,2z-4-4-ethoxyphenyl amino-4-oxobut-2-enoic acid,n-4-ethoxyphenyl maleamic acid,2z-3-n-4-ethoxyphenyl carbamoyl prop-2-enoic acid,2z-3-4-ethoxyphenyl carbamoyl prop-2-enoic acid,4-4-ethoxyanilino-4-oxobut-2-enoic acid,z-4-4-ethoxyanilino-4-oxobut-2-enoic acid,2-butenoic acid,4-4-ethoxyphenyl amino-4-oxo-, 2z
1808883
(Z)-4-(4-ethoxyanilino)-4-oxobut-2-enoic acid
CCOC1=CC=C(C=C1)NC(=O)C=CC(=O)O
Melting Point 188°C to 189°C
Quantity 25g
Formula Weight 235.24
Percent Purity 97%
Chemical Name or Material N-(4-Ethoxyphenyl)maleamic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures