Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-(4-Fluorobenzyl)-3-nitro-p-phenylenediamine, 98%

Click to view available options
Quantity:
1g
5g
150812-21-8
C13H12FN3O2
261.256
MFCD12828054
XTDZJOIEYRRRGJ-UHFFFAOYSA-N
n1-4-fluorobenzyl-3-nitrobenzene-1,4-diamine,1-n-4-fluorophenyl methyl-3-nitrobenzene-1,4-diamine,2-amino-5-4-fluorobenzyl amino-1-nitrobenzene,fluorophenylmethylnitrobenzenediamine,n1-4-fluorophenyl methyl-3-nitrobenzene-1,4-diamine,4-4-fluorobenzylamino-2-nitroaniline,n-4-fluorobenzyl-3-nitro-p-phenylenediamine,n1-4-fluorobenzyl-3-nitro-1,4-phenylenediamine,n~4~-4-fluorophenyl methyl-2-nitrobenzene-1,4-diamine,n4-4-fluorophenyl methyl-2-nitro-benzene-1,4-diamine
20096587
4-N-[(4-fluorophenyl)methyl]-2-nitrobenzene-1,4-diamine
C1=CC(=CC=C1CNC2=CC(=C(C=C2)N)[N+](=O)[O-])F
150812-21-8
C13H12FN3O2
261.256
MFCD12828054
XTDZJOIEYRRRGJ-UHFFFAOYSA-N
n1-4-fluorobenzyl-3-nitrobenzene-1,4-diamine,1-n-4-fluorophenyl methyl-3-nitrobenzene-1,4-diamine,2-amino-5-4-fluorobenzyl amino-1-nitrobenzene,fluorophenylmethylnitrobenzenediamine,n1-4-fluorophenyl methyl-3-nitrobenzene-1,4-diamine,4-4-fluorobenzylamino-2-nitroaniline,n-4-fluorobenzyl-3-nitro-p-phenylenediamine,n1-4-fluorobenzyl-3-nitro-1,4-phenylenediamine,n~4~-4-fluorophenyl methyl-2-nitrobenzene-1,4-diamine,n4-4-fluorophenyl methyl-2-nitro-benzene-1,4-diamine
20096587
4-N-[(4-fluorophenyl)methyl]-2-nitrobenzene-1,4-diamine
C1=CC(=CC=C1CNC2=CC(=C(C=C2)N)[N+](=O)[O-])F
Quantity 5g
Color Brown to Red
Beilstein 22723917
Formula Weight 261.26
Physical Form Powder
Percent Purity 98%
Chemical Name or Material N-(4-Fluorobenzyl)-3-nitro-p-phenylenediamine

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger