Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(4-Hydroxyphenyl)glycine 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
Plastic bottle
122-87-2
C8H9NO3
167.164
MFCD00002689
WRUZLCLJULHLEY-UHFFFAOYSA-N
n-4-hydroxyphenyl glycine,photoglycine,p-hydroxyphenylglycine,hydroxyphenyl glycine,p-hydroxyanilinoacetic acid,iconyl,p-hydroxyphenyl glycine,glycine, n-4-hydroxyphenyl,4-carboxymethylamino phenol,glitsin
67149
CHEBI:55443
2-(4-hydroxyanilino)acetic acid
C1=CC(=CC=C1NCC(=O)O)O
122-87-2
C8H9NO3
167.164
MFCD00002689
WRUZLCLJULHLEY-UHFFFAOYSA-N
n-4-hydroxyphenyl glycine,photoglycine,p-hydroxyphenylglycine,hydroxyphenyl glycine,p-hydroxyanilinoacetic acid,iconyl,p-hydroxyphenyl glycine,glycine, n-4-hydroxyphenyl,4-carboxymethylamino phenol,glitsin
67149
CHEBI:55443
2-(4-hydroxyanilino)acetic acid
C1=CC(=CC=C1NCC(=O)O)O
Infrared Spectrum Authentic
Packaging Plastic bottle
Melting Point 248.0°C
Loss on Drying 1% max.
Quantity 25g
Color Beige-Brown to White or Brown-Gray
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula HOC6H4NHCH2CO2H
Beilstein 13, 488
Merck Index 15, 4875
Formula Weight 167.16
Physical Form Powder
Percent Purity 99%
Chemical Name or Material N-(4-Hydroxyphenyl)glycine, 99%
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 204-580-8

TSCA : TSCA