Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(4-Hydroxyphenylacetyl)spermine, Tocris Bioscience™

Click to view available options
Quantity:
250μg
This item is not returnable. View return policy
130210-32-1
C18H32N4O2
336.48
DRHUIJOIIKXHEH-UHFFFAOYSA-N
n-4-hydroxyphenylacetyl-spermine,n-4-hydroxyphenylacetyl spermine,n-3-4-3-aminopropyl amino butyl amino propyl-2-4-hydroxyphenyl acetamide,n-n-4-hydroxyphenylacetyl-3-aminopropyl-n'-3-aminopropyl-1,4-butanediamine
14622098
N-[3-[4-(3-aminopropylamino)butylamino]propyl]-2-(4-hydroxyphenyl)acetamide
C1=CC(=CC=C1CC(=O)NCCCNCCCCNCCCN)O
This item is not returnable. View return policy
130210-32-1
C18H32N4O2
336.48
DRHUIJOIIKXHEH-UHFFFAOYSA-N
n-4-hydroxyphenylacetyl-spermine,n-4-hydroxyphenylacetyl spermine,n-3-4-3-aminopropyl amino butyl amino propyl-2-4-hydroxyphenyl acetamide,n-n-4-hydroxyphenylacetyl-3-aminopropyl-n'-3-aminopropyl-1,4-butanediamine
14622098
N-[3-[4-(3-aminopropylamino)butylamino]propyl]-2-(4-hydroxyphenyl)acetamide
C1=CC(=CC=C1CC(=O)NCCCNCCCCNCCCN)O

Wasp toxin analog

Wasp toxin analog. More potent than PhTX-343 at locust NMJ. Reported to be selective for NMDA receptors in mammalian CNS.
Quantity 250μg
Formula Weight 336.48
Percent Purity >98%
Chemical Name or Material N-(4-Hydroxyphenylacetyl)spermine

Recommended Storage : Desiccate at -20°C