Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-(4-Iodophenyl)maleamic acid, 97%

Click to view available options
Quantity:
1g
5g
25g
17280-49-8
C10H8INO3
317.082
MFCD00020473
KSVGHDGDESFZOK-WAYWQWQTSA-N
n-4-iodophenyl maleamic acid,2z-3-4-iodophenyl carbamoyl prop-2-enoic acid,4-4-iodoanilino-4-oxobut-2-enoic acid,z-4-4-iodoanilino-4-oxobut-2-enoic acid,2z-3-n-4-iodophenyl carbamoyl prop-2-enoic acid
1549960
(Z)-4-(4-iodoanilino)-4-oxobut-2-enoic acid
C1=CC(=CC=C1NC(=O)C=CC(=O)O)I
17280-49-8
C10H8INO3
317.082
MFCD00020473
KSVGHDGDESFZOK-WAYWQWQTSA-N
n-4-iodophenyl maleamic acid,2z-3-4-iodophenyl carbamoyl prop-2-enoic acid,4-4-iodoanilino-4-oxobut-2-enoic acid,z-4-4-iodoanilino-4-oxobut-2-enoic acid,2z-3-n-4-iodophenyl carbamoyl prop-2-enoic acid
1549960
(Z)-4-(4-iodoanilino)-4-oxobut-2-enoic acid
C1=CC(=CC=C1NC(=O)C=CC(=O)O)I
Quantity 1g
Melting Point 193°C to 194°C
Formula Weight 317.08
Percent Purity 97%
Chemical Name or Material N-(4-Iodophenyl)maleamic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning