Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(4-Nitrobenzoyl)-L-glutamic acid hemihydrate, 98%, Thermo Scientific™

Catalog Number AC205760050
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
6758-40-3
0·5 H2O
305.25
MFCD00149508
NOJZBJAFCSWMKC-VIFPVBQESA-N
s-2-4-nitrobenzamido pentanedioic acid, p-nitrobenzoyl-l-glutamic acid, l-glutamic acid, n-4-nitrobenzoyl, n-p-nitrobenzoyl-l-glutamic acid, n-4-nitrobenzoyl-l-glutamic acid, n-4-nitrobenzoyl-l-glutamic acid hemihydrate, 2s-2-4-nitrophenyl formamido pentanedioic acid, glutamic acid, n-p-nitrobenzoyl-, l, n-4-nitrobenzoyl glutamic acid #
2724179
C1=CC(=CC=C1C(=O)NC(CCC(=O)O)C(=O)O)[N+](=O)[O-]
6758-40-3
0·5 H2O
305.25
MFCD00149508
NOJZBJAFCSWMKC-VIFPVBQESA-N
s-2-4-nitrobenzamido pentanedioic acid, p-nitrobenzoyl-l-glutamic acid, l-glutamic acid, n-4-nitrobenzoyl, n-p-nitrobenzoyl-l-glutamic acid, n-4-nitrobenzoyl-l-glutamic acid, n-4-nitrobenzoyl-l-glutamic acid hemihydrate, 2s-2-4-nitrophenyl formamido pentanedioic acid, glutamic acid, n-p-nitrobenzoyl-, l, n-4-nitrobenzoyl glutamic acid #
2724179
C1=CC(=CC=C1C(=O)NC(CCC(=O)O)C(=O)O)[N+](=O)[O-]

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 111°C to 118°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula HO2CCH2CH2CH(NHCOC6H4NO2)CO2H·1/2H2O
Quantity 5 g
Beilstein 09, III, 1737
Specific Rotation Condition +15.00 (20.00°C c=2,0.1N NaOH)
Specific Rotation + 15
Formula Weight 305.25
Percent Purity 98%
Physical Form Powder
Chemical Name or Material N-(4-Nitrobenzoyl)-L-glutamic acid hemihydrate, 98%
Show More Show Less
EINECSNumber 229-820-9
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit